missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Plastic Pulley Collection
A collection of pulleys for use in classroom demonstrations.
$14.50 - $32.82
Specifications
| Material | Plastic, Metal |
|---|---|
| Capacity (English) | 30 lb. |
| For Use With (Application) | Classroom Demonstrations |
| Includes | 2 Hooks |
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
|
S99351
|
Eisco™
PH0280A |
Single Pulley |
Each for $14.50
|
|
|||||
|
S99352
|
Eisco™
PH0280B |
Double Parallel Pulley |
Each for $17.25
|
|
|||||
|
S15117
|
Eisco™
PH0280D |
Double Long Pulley |
Each for $32.82
|
|
|||||
|
S99353
|
Eisco™
PH0280C |
Triple Parallel Pulley |
Each for $18.11
|
|
|||||
Description
- High quality plastic pulleys mounted in steel frame
- 2 hooks - 1 attached to each end of the steel mounting for hanging
- Capable of holding up to 30 lb. of weight
Specifications
| Plastic, Metal | |
| 30 lb. | |
| Classroom Demonstrations | |
| 2 Hooks |