missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco™ Plastic Pulley Collection
A collection of pulleys for use in classroom demonstrations.
Supplier: Eisco™ PH0280D
Description
- High quality plastic pulleys mounted in steel frame
- 2 hooks - 1 attached to each end of the steel mounting for hanging
- Capable of holding up to 30 lb. of weight
Identifiants chimiques
| Plastic, Metal | |
| 30 lb. | |
| Classroom Demonstrations | |
| 2 Hooks |
Spécifications
| Double Long Pulley | |
| 30 lb. | |
| Classroom Demonstrations | |
| 2 Hooks | |
| 11.4 cm |
| Plastic, Metal | |
| 1.5 in., 2 in. | |
| 3.8 cm, 5 cm | |
| 4.5 in. |