missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Sulfo-NHS-Acetate

Catalog No. PI26777
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI26777 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI26777 Supplier Thermo Scientific™ Supplier No. 26777

missing translation for 'validMainframeMsg'

Amine-reactive (NHS ester) compound to permanently block primary amines (e.g., protein lysine side chains) to prevent interactions or later conjugation.

Thermo Scientific™ Pierce Sulfo-NHS-Acetate is a small compound that forms stable, covalent amide bonds with primary amines (e.g., lysine side chains), enabling them to be permanently blocked to prevent interactions or later conjugation.

Highlights:

  • Reacts mainly with primary amines (lysine ε amino groups)
  • Irreversible reaction is accompanied by the release of N-hydroxysulfosuccinimide

Specifications

Chemical Reactivity Amine
Content And Storage Upon receipt store desiccated at -20° C. Product is shipped at ambient temperature.
Label or Dye Unreactive Group
Label Type Blocking Groups
Product Line Pierce
Reactive Moiety Active Ester, Succinimidyl Ester, NHS Ester
Spacer Short (<10 Å)
Form Powder
Quantity 100 mg
Solubility DMSO, Water, DMF
Product Type Sulfo-NHS-Acetate
Show More Show Less
Can you provide the shelf-life for Sulfo-NHS-Acetate?

Sulfo-NHS-Acetate is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended (-20 degrees C). Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.

For Research Use Only. Not for use in diagnostic procedures.