missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Diethanolamine Substrate Buffer (5X)

Catalog No. PI34064
Change view
Click to view available options
Quantity:
225 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI34064 225 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI34064 Supplier Thermo Scientific™ Supplier No. 34064

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimized, reliable and convenient stock solutions of peroxide for HRP-based assays and diethanolamine for alkaline phosphatase (alk-phos) detection.

Thermo Scientific™ Pierce Substrate Buffers include optimized, reliable and convenient stock solutions of peroxide for HRP-based assays and diethanolamine for alkaline phosphatase (alk-phos) detection.

The Stable Peroxide Substrate Buffer is an ideal formulation for researchers who prefer to make their own horseradish peroxidase (HRP) substrates. The substrate is easily prepared by adding the chosen chromogen to a 1X solution of the Stable Peroxide Substrate Buffer, which supplies the needed peroxide and buffer condition to obtain optimum and consistent results. This buffer is provided at a 10X concentrate, which allows a shelf life of up to one year. The Diethanolamine Buffer is a 5X stock solution (5.1M, pH 9.8) that provides optimal conditions for development of alkaline phosphatase reactions, such as with PNPP (Part No. 34045, 34047).

Highlights:

  • Ready-to-use concentrated stock solutions
  • Minimize assay-to-assay variability

Stable Peroxide Substrate Buffer (10X) (Part No. 34062)

  • Long shelf life: 12 months after receipt
  • Total volume of 1000mL can be made from one 100mL bottle of concentrate
  • Add chromogen to stable peroxide substrate buffer for detection of horseradish peroxidase

Diethanolamine Substrate Buffer (5X) (Part No. 34064)

  • For use with soluble alkaline phosphatase substrates such as PNPP
  • Optimized, consistent pH
  • Stable even at a 1X concentration

Specifications

Description Pierce Diethanolamine Substrate Buffer, 5X
Product Type ELISA Substrate
Quantity 225 mL
Form Liquid
For Use With (Application) ELISA, Immunohistochemistry (IHC), Western Blot
Product Line Pierce
Substrate Type Alkaline Phosphatase, Horseradish Peroxidase
Content And Storage Upon receipt store product at 4°C. Product is shipped at ambient temperature.

For Research Use Only. Not for use in diagnostic procedures.