Learn More
Thermo Scientific™ Pierce™ Protein-Free Blocking Buffer
Pierce Protein-Free Blocking Buffer is a non-protein compound that provides effective blocking for membrane-based and plate-based protein detection methods, resulting in extremely low background.
Supplier: Thermo Scientific™ 37573
Description
Pierce Protein-Free Blocking Buffer is a non-protein compound that provides effective blocking for membrane-based and plate-based protein detection methods, resulting in extremely low background. It is supplied in a convenient, ready-to-use format.
Features of Protein-Free Blocking Buffer include:
- Protein-free blocker—minimizes or eliminates cross-reactivity associated with protein-based blocking buffers
- Applications—compatible with western blotting (membranes), ELISA (microplates), and arrays (coated glass slides)
- Streptavidin-friendly—free of biotin; no interference with avidin-biotin detection systems
- High-performance—optimized and validated in many protein methods to provide high signal-to-noise ratio (i.e., no quenching of specific binding and signal but eliminating nonspecific binding and background)
Traditional blocking buffers contain proteins that can cross-react in certain immunodetection systems, resulting in high background and reduced signal. Pierce Protein-Free Blocking Buffers are devoid of protein while remaining highly effective at blocking plates, membranes, and other surfaces for ELISA, western blotting, glass slide arrays, and other applications.
Identifiants chimiques
| 1X | |
| ELISA | |
| Liquid |
Spécifications
| Protein-Free T20 (PBS) | |
| ELISA | |
| Pierce |
| 1X | |
| Liquid | |
| 1 L |
Sécurité et manipulation
Recommended Storage : Upon receipt store product at room temperature. After opening, store product at 4°C.