missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Angelicin, ≥98% (HPLC), MilliporeSigma™ Supelco™

Numéro de catalogue. 111002018
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111002018 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111002018 Fournisseur Merck Emd Millipore Code fournisseur PHL8987410MG
Il en reste null

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Spécifications

CAS 523-50-2
Product Line phyproof
Linear Formula C11H6O3
MDL Number MFCD00064930
Synonym 2-Oxo-(2 H)-furo(2,3-h)-1-benzopyran; 2 H-Furo[2,3-h]-1-benzopyran-2-one; Isopsoralen; NSC 404563
Type Reference Substance
Quantity 10 mg
Formula Weight 186.16
Percent Purity ≥98% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG