missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphate Buffer pH 7.2, For BOD, Certified, LabChem™
$58.07 - $181.82
Specifications
| Chemical Name or Material | Phosphate Buffer pH 7.2 |
|---|---|
| Name Note | For BOD |
| CAS Max % | 95.03,2.18,0.85,1.77,0.17 |
| Color | Colorless |
| CAS | 7758-11-4 , 7778-77-0 , 7558-79-4 , 12125-02-9 |
Specifications
| Phosphate Buffer pH 7.2 | |
| 95.03,2.18,0.85,1.77,0.17 | |
| 7758-11-4 , 7778-77-0 , 7558-79-4 , 12125-02-9 | |
| 1g/mL | |
| Liquid | |
| KH2PO4 | |
| 1g/mL | |
| Soluble in water | |
| Certified |
| For BOD | |
| Colorless | |
| Gaseous Ammonia; Phosphorus oxides | |
| 7.2 ±0.1 | |
| Passes Test | |
| Poly Bottle | |
| Traceable to NIST | |
| 136.09 |