Learn More
Phosphate Buffer pH 7.2, For BOD, Certified, LabChem™
Supplier:  LabChem LC185002
 View more versions of this product
Catalog No. LC185002
Only null left
Description
Description
Specifications
Specifications
Phosphate Buffer pH 7.2
95.03,2.18,0.85,1.77,0.17
7758-11-4 , 7778-77-0 , 7558-79-4 , 12125-02-9
1g/mL
Liquid
Passes Test
Poly Bottle
Traceable to NIST
136.09
For BOD
Colorless
Gaseous Ammonia; Phosphorus oxides
7.2 ±0.1
1 L
KH2PO4
1g/mL
Soluble in water
Certified
Product Suggestions
Product Suggestions