missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phire Reaction Buffers

Numéro de catalogue. F525L
Change view
Click to view available options
Chemical Name or Material:
Phire Green Reaction Buffer
Phire Reaction Buffer
Phire Reaction Buffer, detergent-free
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Chemical Name or Material
F525L Phire Reaction Buffer, detergent-free
F524L Phire Reaction Buffer
F527L Phire Green Reaction Buffer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Numéro de catalogue. F525L Fournisseur Thermo Scientific™ Code fournisseur F525L
Il en reste null

The reaction buffer for Thermo Scientific Phire Hot Start II DNA Polymerase is available in three formats: non-colored version with and without detergents or Green version containing a density reagent and two tracking dyes for direct loading of PCR products on gel.

Thermo Scientific Phire Reaction Buffer is designed for Phire Hot Start II DNA Polymerase. This 5X buffer solution is available in three formats, a non-colored version with or without detergents and a green version that contains a density reagent and two tracking dyes for direct loading of PCR products on gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

Phire Reaction Green Buffer includes a blue die that migrates with 3–5 kb DNA fragments in 1% agarose gel and a yellow dye that migrates faster than 10 bp DNA fragments in 1% agarose gel. The dyes have excitation peaks at 424 nm and 615 nm.

The concentration of Mg2+ in Phire Reaction Buffer is optimized to work well for most amplicons. The buffer contains 1.5 mM MgCl2 at final reaction concentration. Separate tubes of MgCl2 and DMSO are provided for further optimization.

Notes

  • For applications that require PCR product analysis by absorbance or fluorescence excitation, the colorless Phire Reaction Buffer is recommended.

Spécifications

Chemical Name or Material Phire Reaction Buffer, detergent-free
Description Detergent-free
Concentration 5X
For Use With (Application) Standard PCR
Quantity 4 x 1.5 mL
Includes • 5X Phire Reaction Buffer, detergent-free, 4 x 1.5 mL
• 50 mM MgCl2, 2 x 1.5 mL
• 100% DMSO, 0.5 mL

Store at -20°C.

For Research Use Only. Not for use in diagnostic procedures.