missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Phire Hot Start II PCR Master Mixes

Thermo Scientific Phire Hot Start II PCR Master Mix is convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phire Hot Start II DNA Polymerase, nucleotides and optimized reaction buffer including MgCl2.

Supplier:  Thermo Scientific™ F126L

 Afficher plus de versions de ce produit

Numéro de catalogue. F126L


Il en reste null

Comprend: Phire Hot Start II DNA Polymerase is supplied with 5x Phire Reaction Buffer and DMSO. The buffer provides 1.5 mM MgCl2 in the final 1x concentration.

Description

Description

Thermo Scientific Phire Hot Start II PCR Master Mix is a convenient 2X mix designed to minimize the number of pipetting steps. The master mix contains Phire Hot Start II DNA Polymerase, nucleotides, and optimized reaction buffer, including MgCl2. Only template and primers need to be added to the PCR reaction.

Phire Green Hot Start II PCR Master Mix also includes a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with enzyme performance and is compatible with downstream applications such as DNA sequencing, ligation, and restriction digestion.

Phire Hot Start II DNA Polymerase, included in the master mix, is an enhanced PCR enzyme for routine and high throughput PCR applications. It outperforms every Taq-based hot start polymerase on the market. Phire Hot Start II DNA Polymerase is significantly faster, extremely robust, and also capable of amplifying long DNA fragments with high yields. These features are achieved through advanced protein engineering of the polymerase. It incorporates a unique double-stranded DNA binding domain that allows short extension times (10–15 s/kb), improves yields, and increases fidelity 2-fold compared to Taq DNA polymerase (Cat. No. EP0072).

Features

  • No reactivation step with quick hot start
  • Amplify four times faster than with hot start Taq
  • Minimal reaction optimization due to high inhibitor tolerance
  • Abundant products due to high efficiency
  • Amplify significantly longer DNA fragments than with any hot start Taq
  • Direct loading on gel with Green format

Applications

  • Hot Start PCR
  • Routine PCR
  • Non-high-fidelity PCR
  • Fast PCR
  • High throughput PCR
  • Genotyping

Notes

  • The optimal annealing temperature for Phire DNA Polymerases may differ significantly from that of Taq-based polymerases.
Spécifications

Identifiants chimiques

2X
Phire Hot Start II DNA Polymerase
Hot Start PCR Master Mix
Cloning, Hot-start PCR
Built-In Hot Start
SuperMix or Master Mix
Medium
Fast
Wet or Dry Ice
2X
Blunt
DNA

Spécifications

2X
Medium
Fast
1,000 reactions
Cloning, Hot-start PCR
Built-In Hot Start
Blunt
DNA
• 2X Phire Green Hot Start II PCR Master Mix, 20 x 1.25 mL
• DMSO, 4 x 0.5 mL
• Nuclease-free water, 20 x 1.25 mL

Store at -20°C.
Phire Hot Start II DNA Polymerase
Hot Start PCR Master Mix
Wet or Dry Ice
2X
1000 Reactions
SuperMix or Master Mix
Green
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.