missing translation for 'onlineSavingsMsg'
Learn More

Phenylarsine Oxide, For BOD, Certified, 0.0375N ±0.0005N (0.01875M), LabChem™

Catalog No. LC183902
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC183902 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC183902 Supplier LabChem Supplier No. LC183902

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 637-03-6 , 7732-18-5 , 7647-01-0 , 1310-73-2
Molecular Formula C6H5AsO
Molecular Weight (g/mol) 168.03
MDL Number MFCD00001990
InChI Key BQVCCPGCDUSGOE-UHFFFAOYSA-N
Synonym phenylarsine oxide, oxophenylarsine, arzene, phenylarsenoxide, arsenosobenzene, arsine, oxophenyl, phenyl arsine oxide, benzene, arsenoso, fenylarsinoxid, caswell no. 060
PubChem CID 4778  
ChEBI CHEBI:75253
IUPAC Name arsorosobenzene
SMILES O=[As]C1=CC=CC=C1

Specifications

Color Colorless to Yellow
CAS Max % 0.33, 98.92, 0.4, 0.35
Linear Formula C6H5AsO
Concentration 0.0375N ±0.0005N (0.01875M)
Solubility Information Soluble in water
Formula Weight 168.03
Grade Certified
Identification Passes Test
Packaging Amber Glass
Traceability to NIST Traceable to NIST
Decomposition Information Arsenic and its oxides; Carbon monoxide; Carbon dioxide
pH 7.0 to 9.0
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Phenylarsine Oxide, For BOD
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear eye protection, protective gloves.
Wash exposed skin thoroughly after handling.
If on skin: Wash with plenty of soap and water.
Take off contaminated clothing.
If skin irritation occurs: Get medical advice/attention.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Recommended Storage Room Temperature