missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Phenolic Screw Cap with Open Center

Phenolic screw cap with open center

$54.80 - $96.40

Specifications

Color Black
No. per Pack 100
Product Type Phenolic Hole Cap
View More Specs

Products 3
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
111011497
View Documents Promotion Details
MilliporeSigma™ Supelco™
27176
22 mL Vial
Each for $95.60
Only null left
 
111011498
View Documents Promotion Details
MilliporeSigma™ Supelco™
SGMA27187
20 or 40 mL Vial
Each for $96.40
Only null left
 
111011884
View Documents Promotion Details
MilliporeSigma™ Supelco™
27154
7 mL Vial
Each for $54.80
Only null left
 
Description

Description

  • Screw caps are available as hole caps for use with sealing septa.
  • Caps are offered in phenolic resin.
  • Phenolic hole caps can withstand a maximum temperature of 149°C.
Specifications

Specifications

Black
100
Phenolic Hole Cap
Videos
SDS
Documents

Documents