missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Phenolic Screw Cap with Open Center

Phenolic screw cap with open center

Supplier:  MilliporeSigma™ Supelco™ SGMA27187

 View more versions of this product

Catalog No. 111011498


Only null left

Description

Description

  • Screw caps are available as hole caps for use with sealing septa.
  • Caps are offered in phenolic resin.
  • Phenolic hole caps can withstand a maximum temperature of 149°C.
Specifications

Chemical Identifiers

Black
100
Phenolic Hole Cap

Specifications

Black
24-400
Phenolic Hole Cap
100
20 or 40 mL Vial
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions