missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Phenolic Screw Cap with Open Center

Catalog No. 111011497
Change view
Click to view available options
For Use With (Equipment):
20 or 40 mL Vial
22 mL Vial
7 mL Vial
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
111011497 22 mL Vial
111011498 20 or 40 mL Vial
111011884 7 mL Vial
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111011497 Supplier MilliporeSigma™ Supelco™ Supplier No. 27176
Only null left

Phenolic screw cap with open center

  • Screw caps are available as hole caps for use with sealing septa.
  • Caps are offered in phenolic resin.
  • Phenolic hole caps can withstand a maximum temperature of 149°C.

Specifications

Color Black
No. per Pack 100
Closure Size 20-400
For Use With (Equipment) 22 mL Vial
Product Type Phenolic Hole Cap