missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Vernolat, MilliporeSigma™ Supelco™
Description
Agriculture
Spécifications
Spécifications
| Product Line | PESTANAL |
| CAS | 1929-77-7 |
| Physical Form | Neat |
| Linear Formula | C10H21 NOS |
| MDL Number | MFCD00078733 |
| Synonym | S-Propyl dipropylthiocarbamate |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 203.34 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH