missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Metazachlor, MilliporeSigma™ Supelco™
Description
Agriculture
Spécifications
Spécifications
| Product Line | PESTANAL |
| CAS | 67129-08-2 |
| Physical Form | Neat |
| Linear Formula | C14H16 ClN3 O |
| MDL Number | MFCD00055509 |
| Melting Point | 74°C to 78°C |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 277.75 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH