missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Mephosfolan, MilliporeSigma™ Supelco™
Description
Agriculture
Spécifications
Spécifications
| Product Line | PESTANAL |
| CAS | 950-10-7 |
| Physical Form | Neat |
| Linear Formula | C8H16 NO3 PS2 |
| MDL Number | MFCD00128050 |
| Synonym | Diethyl (4-methyl-1,3-dithiolan-2-ylidene)phosphoramidate |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 269.32 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH