missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Cycluron, MilliporeSigma™ Supelco™

Numéro de catalogue. 111013545
Modifier l'affichage
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111013545 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111013545 Fournisseur Merck Emd Millipore Code fournisseur 45409250MG
Il en reste null
Agriculture

Spécifications

Product Line PESTANAL
CAS 2163-69-1
Flash Point >100°C
Physical Form Neat
Linear Formula C11H22N2 O
MDL Number MFCD00055376
Synonym 3-Cyclooctyl-1,1-dimethylurea
Type Analytical Standard
Quantity 250 mg
Formula Weight 198.31
Grade Analytical Standard
Afficher plus de résultats Afficher moins de résultats

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH