missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PESTANAL™ Bixafen, MilliporeSigma™ Supelco™
Description
Agriculture
Spécifications
Spécifications
| Product Line | PESTANAL |
| CAS | 581809-46-3 |
| Physical Form | Neat |
| Linear Formula | C18H12 Cl2F3N3 O |
| MDL Number | MFCD16652583 |
| Synonym | N-(3 ′,4 ′-Dichloro-5-fluorobiphenyl-2-yl)-3-(difluoromethyl)-1-methylpyrazole-4-carboxamide |
| Type | Analytical Standard |
| Quantity | 100 mg |
| Formula Weight | 414.21 |
| Grade | Analytical Standard |
PESTANAL is a registered trademark of Sigma-Aldrich International GmbH