missing translation for 'onlineSavingsMsg'
Learn More

PESTANAL™ Abamectin, MilliporeSigma™ Supelco™

Numéro de catalogue. 111019256
Modifier l'affichage
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
111019256 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 111019256 Fournisseur Merck Emd Millipore Code fournisseur 31732100MG
Il en reste null

Abamectin (MK-0936) belongs to avermectin family. This family have a macrocycle as backbone with a spiroketal unit, a hexahydrobenzofuran unit and disaccharide substitute in general. MK-0936 is a natural fermenting product which can be produced by Streptomyces avermitilis.

Agriculture and Environmental

Spécifications

Product Line PESTANAL
CAS 71751-41-2
Physical Form Neat
MDL Number MFCD01769550
Synonym Avermectin B1
Type Analytical Standard
Quantity 100 mg
Grade Analytical Standard

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH