missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pellino 3 Monoclonal Antibody (K40052_17C1), Invitrogen™
Mouse Monoclonal Antibody
$1,461.54 - $2,194.06
Spécifications
| Antigen | Pellino 3 |
|---|---|
| Clone | K40052_17C1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Numéro de catalogue | Numéro du manufacturier. | Quantity | Prix | Quantity & Availability | |||||
|
MA556159
|
Invitrogen™
MA556159 |
200 μL |
chaque for $2,194.06
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
|
MA555779
|
Invitrogen™
MA555779 |
50 μL |
chaque for $1,461.54
|
|
Connectez-vous ou enregistrez-vous pour vérifier votre prix et la disponibilité.
|
||||
Description
Sequence of this protein is as follows: SPRTSDLQHR GNKGSCVLSS PGEDAQPGEE PIKYGELIVL GCCEEGGEET EAQRGEVTGP RAHSCYNGCL ASGDKGRRRS RLALSRRSHA NGVKPDVMHH ISTPLVSKAL SNRGQHSISY TLSRSHSVIV EYTHDSDTDM FQIGRSTENM IDFVVTDTSP GGGAAEGPSA
Pellino 3 Monoclonal antibody specifically detects Pellino 3 in Human samples. It is validated for Western BlotSpécifications
| Pellino 3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 6030441F14Rik; A930011L17; BC028931; E3 ubiquitin-protein ligase pellino homolog 3; E3 ubiquitin-protein ligase pellino homolog 3 pseudogene; MGC35521; PELI3; pellino 3; pellino E3 ubiquitin protein ligase family member 3; pellino homolog 3; pellino-3; protein pellino homolog 3; RGD1305989; RING-type E3 ubiquitin transferase pellino homolog 3 | |
| Recombinant human PELI3 (immunogen range is 11-180). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PELI3 |
| K40052_17C1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8N2H9 | |
| 246330 | |
| IgG1 | |
| Protein A | |
| Antibody |