missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Pellino 3 Monoclonal Antibody (K40052_17C1), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA556159
Description
Sequence of this protein is as follows: SPRTSDLQHR GNKGSCVLSS PGEDAQPGEE PIKYGELIVL GCCEEGGEET EAQRGEVTGP RAHSCYNGCL ASGDKGRRRS RLALSRRSHA NGVKPDVMHH ISTPLVSKAL SNRGQHSISY TLSRSHSVIV EYTHDSDTDM FQIGRSTENM IDFVVTDTSP GGGAAEGPSA
Pellino 1 is the mammalian homologue of Pellino, a Drosophila Pelle-interacting protein. Pelle is a serine-threonine kinase through which Toll signaling can occur. Pellino 1 can activate c-Jun, Elk-1 and NF-kB, whereas Pellino 3 can only activate c-Jun and Elk-1(2).Chemical Identifiers
| Pellino 3 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 6030441F14Rik; A930011L17; BC028931; E3 ubiquitin-protein ligase pellino homolog 3; E3 ubiquitin-protein ligase pellino homolog 3 pseudogene; MGC35521; PELI3; pellino 3; pellino E3 ubiquitin protein ligase family member 3; pellino homolog 3; pellino-3; protein pellino homolog 3; RGD1305989; RING-type E3 ubiquitin transferase pellino homolog 3 | |
| Recombinant human PELI3 (immunogen range is 11-180). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| PELI3 |
| K40052_17C1 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q8N2H9 | |
| 246330 | |
| IgG1 | |
| Protein A | |
| Antibody |
Specifications
| Pellino 3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q8N2H9 | |
| PELI3 | |
| Recombinant human PELI3 (immunogen range is 11-180). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K40052_17C1 | |
| Unconjugated | |
| PELI3 | |
| 6030441F14Rik; A930011L17; BC028931; E3 ubiquitin-protein ligase pellino homolog 3; E3 ubiquitin-protein ligase pellino homolog 3 pseudogene; MGC35521; PELI3; pellino 3; pellino E3 ubiquitin protein ligase family member 3; pellino homolog 3; pellino-3; protein pellino homolog 3; RGD1305989; RING-type E3 ubiquitin transferase pellino homolog 3 | |
| Mouse | |
| Protein A | |
| RUO | |
| 246330 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |