Learn More
Cytiva PEEK Finger Tight Connector
Catalog No. 361041172
Change view
Click to view available options
Connections:
1/16 in. Male
Catalog No. Connections
361041172 1/16 in. Male
1 options
Catalog No. 361041172 Supplier Cytiva Supplier No. 18111255
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Use With Tubing o.d. 1/16 in. Cytiva PEEK Finger Tight Connector provides the tightest connections.
  • Packsize 10 Connections
  • Fingertight 1/16 in.
  • Polyetheretherketone (PEEK) 
Specifications
Connections 1/16 in. Male