Learn More
Thermo Scientific Chemicals Pectinase, Aspergillus niger
Catalog No. AAJ63408MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63408MD 250 mg
AAJ6340803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ63408MD Supplier Thermo Scientific Chemicals Supplier No. J63408MD
Used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation.
Pectinase, Aspergillus niger is used in plant protoplast preparation to digest cell wall prior to organelle isolation. Pectinase helps facilitate the maceration of plant materials and extraction processes including organelle preparation. It is used in the enzyme preparation. Used mostly to reduce viscosity, to improve filtration to clarify products, to avoid particle sedimentation and to prevent pectin gel formation in the manufacture of fruit juice and wine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity 250 mg
Form Powder
RUO – Research Use Only