missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Paraformaldehyde fixative solution

Numéro de catalogue. p-7042286
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
AAJ61984AK 250 mL
AAJ61984AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Numéro de catalogue. AAJ61984AK Fournisseur Thermo Scientific Chemicals Code fournisseur J61984AK
Il en reste null

Paraformaldehyde fixative solution finds application in proteins, peptides and the enzymes of low molecular weight.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Physical Form Liquid
Chemical Name or Material Paraformaldehyde fixative solution
Quantity 250 mL
MDL Number MFCD00133991
Solubility Information Miscible with water.

RUO – Research Use Only