Learn More
Motic™ Panthera TEC-Epi Polarizing Trinocular Microscope Package
Numéro de catalogue. 01257461
Change view
Click to view available options
Camera Type:
6 MP Digital
Numéro de catalogue. Camera Type
01257461 6 MP Digital
1 options
Numéro de catalogue. 01257461 Fournisseur Motic™ Code fournisseur MBPANPETS6
Pour de laide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
The Panthera TEC POL offers a new level of transmitted light microscopes for the investigation of birefringent samples. This package includes the Panthera TEC Polarizing Trinocular Microscope with 0.5X C-mount camera adapter, Moticam and Motic™ Analysis Basic 64-bit.
Even transparent birefringent structures from biology and medicine (heart muscle cells, secondary cell walls, etc.) can be detected by the Full Koehler illumination of Panthera TEC POL Microscopes. With Panthera TEC POL models, now opaque industrial samples can be examined to detect bireflection of flat polished and etched surfaces.
  • Interchangeable 30 W Halogen/3 W LED light source is part of full Koehler setup with field and aperture diaphragm
  • Built-in low energy mode and LED feedback indicator helps optimize workflow
  • LED incident light source can easily be changed to different color temperatures (3500 K or 5500 K)
  • UC Plan Achromatic objectives focus on strain-free mounting for maximum darkness of image background
  • 5-fold encoded nosepiece memorizes light intensity for each objective position to replicate illumination once objective is swung in again
  • Intermediate tube with focusable and centerable Bertrand lens is prepared for conoscopic analysis of crystals and minerals in transmitted light
  • Extended field of view offers 21% more visual area in comparison to basic 20 mm system
  • All trinocular versions (25° viewing angle) have fixed beam split of 50/50 (visual/camera port)
  • 360° rotatable stage runs smoothly for easy positioning of sample, bringing regions precisely into crossing point of eyepiece reticle
  • Diopter adjustment
Spécifications
Magnification Power 10X
Objective Types CCIS UC Plan 5X, 10X, 20X, 40X
Head Style Trinocular
Camera Type 6 MP Digital
Lighting Type Kohler Illumination
Eyepiece WF 10X/22 mm