Learn More
Motic Panthera E2 Microscope and Camera Bundle
Designed for the future, the Panthera E2 is ready for ambitious work in educational environments and small life science labs. Features new CCIS™ Plan Ultra Contrast Objectives with stunning image quality in Brightfield, Phase Contrast, Darkfield, and Polarization.
Supplier: Motic 1100403300212
Description
- Swiveling, 25° inclined Siedentopf head
- LED light indicator
- Coaxial coarse and fine focusing adjustment with tension adjustment
- Built-in, low position coaxial rackless stage with right hand control
- Focusable Abbe condenser with 0.9 numerical aperature
- Iris diaphragm and slot li>Diopter Adjustment: ±5
Identifiants chimiques
| Panthera E2 Trinocular | |
| LED |
Spécifications
| BTI10 Camera Bundle | |
| CCIS Plan UC Objectives 4X, 10X, 40X S, 100X S-oil | |
| Trinocular | |
| LED |
| 10X | |
| Panthera E2 Trinocular | |
| Moticam BTI10 | |
| WF 10X/20 mm |