missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Palmitic acid, 98%

CAS: 57-10-3 | C16H32O2 | 256.43 g/mol

Supplier:  Thermo Scientific Chemicals 129700010

Quantity 1 kg
Packaging Plastic bottle

 Afficher plus de versions de ce produit

Numéro de catalogue. AC129700010


Il en reste null

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

57-10-3
256.43
IPCSVZSSVZVIGE-UHFFFAOYSA-N
985
hexadecanoic acid
C16H32O2
MFCD00002747
palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid
CHEBI:15756
CCCCCCCCCCCCCCCC(O)=O

Spécifications

Palmitic acid
57-10-3
White
351.5°C
Authentic
Plastic bottle
CH3(CH2)14CO2H
MFCD00002747
02,IV,1157
palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid
IPCSVZSSVZVIGE-UHFFFAOYSA-N
hexadecanoic acid
985
CHEBI:15756
98%
215 to 233mg KOH/g
59.0°C to 63.0°C
0.8500g/mL
206°C
97.5% min. (GC)
C16H32O2
1 kg
216 to 224mg KOH/g
15,7097
Solubility in water: insoluble. Other solubilities: soluble in hot alcohol, ether and propyl alcohol
CCCCCCCCCCCCCCCC(O)=O
256.43
7.8 mPa.s (70°C)
256.42
Flakes
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

GHS H Statement:
Causes serious eye irritation.
Fatal if swallowed.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 200-312-9

SDS
Documents

Documents

Promotions

Promotions