Learn More
Thermo Scientific Chemicals Palmitic acid, 98%
CAS: 57-10-3 | C16H32O2 | 256.43 g/mol
$72.24 - $1,778.74
Chemical Identifiers
| CAS | 57-10-3 |
|---|---|
| Molecular Formula | C16H32O2 |
| Molecular Weight (g/mol) | 256.43 |
| MDL Number | MFCD00002747 |
| InChI Key | IPCSVZSSVZVIGE-UHFFFAOYSA-N |
| Synonym | palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid |
| PubChem CID | 985 |
| ChEBI | CHEBI:15756 |
| IUPAC Name | hexadecanoic acid |
| SMILES | CCCCCCCCCCCCCCCC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC129702500
|
Thermo Scientific Chemicals
129702500 |
250 g | Plastic bottle |
Each for $72.24
|
|
||||
|
AC129700010
|
Thermo Scientific Chemicals
129700010 |
1 kg | Plastic bottle |
Each for $149.75
|
|
||||
|
AC129700025
|
Thermo Scientific Chemicals
129700025 |
2.5 kg | Plastic bottle |
Each for $336.22
|
|
||||
|
AC129700250
|
Thermo Scientific Chemicals
129700250 |
25 kg | Fibre drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| C16H32O2 | |
| MFCD00002747 | |
| palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid | |
| CHEBI:15756 | |
| CCCCCCCCCCCCCCCC(O)=O |
Specifications
| 215 to 233mg KOH/g | |
| 59.0°C to 63.0°C | |
| 0.8500g/mL | |
| 206°C | |
| 97.5% min. (GC) | |
| CH3(CH2)14CO2H | |
| 216 to 224mg KOH/g | |
| 15,7097 | |
| Solubility in water: insoluble. Other solubilities: soluble in hot alcohol, ether and propyl alcohol | |
| CCCCCCCCCCCCCCCC(O)=O | |
| 256.43 | |
| 7.8 mPa.s (70°C) | |
| 256.42 | |
| Flakes |
| 57-10-3 | |
| White | |
| 351.5°C | |
| Authentic | |
| C16H32O2 | |
| MFCD00002747 | |
| 02,IV,1157 | |
| palmitic acid, cetylic acid, palmitate, n-hexadecanoic acid, hexadecylic acid, hydrofol, n-hexadecoic acid, 1-pentadecanecarboxylic acid, palmitinic acid, pentadecanecarboxylic acid | |
| IPCSVZSSVZVIGE-UHFFFAOYSA-N | |
| hexadecanoic acid | |
| 985 | |
| CHEBI:15756 | |
| 98% | |
| Palmitic acid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Fatal if swallowed.
Causes skin irritation.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
EINECSNumber : 200-312-9
RUO – Research Use Only