missing translation for 'onlineSavingsMsg'
Learn More

Palmatine chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 467150010

 View more versions of this product

Catalog No. AC467150010


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Palmatine chloride
1 g
387.86
Crystalline Powder
≥97.5%
[Cl-].COC1=CC=C2C=C3C4=CC(OC)=C(OC)C=C4CC[N+]3=CC2=C1OC
387.86
10605-02-4
C21 H22 Cl N O4
MFCD00016656
Light-yellow to Yellow to Orange
RLQYRXCUPVKSAW-UHFFFAOYSA-M
3,4,10,11-tetramethoxy-7,8-dihydro-6λ⁵-azatetraphen-6-ylium chloride

Chemical Identifiers

10605-02-4
C21 H22 Cl N O4
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Exclamation mark

Recommended Storage : Normal conditions

SDS
Documents

Documents

Promotions

Promotions