missing translation for 'onlineSavingsMsg'
Learn More

Palmatine chloride, 97%, Thermo Scientific™

Catalog No. p-7238586
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC467152500 250 mg
AC467150010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC467152500 Supplier Thermo Scientific Chemicals Supplier No. 467152500
Only null left

Chemical Identifiers

CAS 10605-02-4
Molecular Formula C21 H22 Cl N O4

Specifications

Quantity 250 mg
Formula Weight 387.86
MDL Number MFCD00016656
Physical Form Crystalline Powder
Color Light-yellow to Yellow to Orange
Purity ≥97.5%
InChI Key RLQYRXCUPVKSAW-UHFFFAOYSA-M
SMILES [Cl-].COC1=CC=C2C=C3C4=CC(OC)=C(OC)C=C4CC[N+]3=CC2=C1OC
IUPAC Name 3,4,10,11-tetramethoxy-7,8-dihydro-6λ⁵-azatetraphen-6-ylium chloride
Molecular Weight (g/mol) 387.86
Chemical Name or Material Palmatine chloride
Show More Show Less
Hazard Category Exclamation mark
Recommended Storage Normal conditions