accessibility menu, dialog, popup

UserName

CAS RN 10605-02-4

IUPAC Name: 3,4,10,11-tetramethoxy-7,8-dihydro-6λ⁵-azatetraphen-6-ylium chloride
Synonyms: palmatine
Molecular Weight (g/mol): 387.86
Molecular Formula: C21H22ClNO4
InChi Key: RLQYRXCUPVKSAW-UHFFFAOYSA-M
SMILES: [Cl-].COC1=CC=C2C=C3C4=CC(OC)=C(OC)C=C4CC[N+]3=CC2=C1OC
Color 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)