missing translation for 'onlineSavingsMsg'
Learn More

Palladium Standard For ICP, 1000μg/mL in 10% HCl, Assurance Grade, SPEX CertiPrep™

Catalog No. p-5533076
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PLPD32Y 125 mL Triple-leached LDPE bottle
PLPD32X 500 mL Triple-leached LDPE bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PLPD32Y Supplier SPEX CertiPrep Group LLC Supplier No. PLPD32Y
Only null left

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Pd1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7440-05-3 , 7647-01-0 , 7732-18-5
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Packaging Triple-leached LDPE bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 39 elements at ppb levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Min % 0.1, 10.0, 89.9
CAS Max % 0.1, 10.0, 89.9
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 10% HCl
Quantity 125 mL
Grade Assurance
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
10% HCl    1000μg/mL    PLPD3-2Y, -2X    10,000    PLPD3-3Y, -3X
Chemical Name or Material Palladium Standard
Show More Show Less
Hazard Category Causes skin irritation, serious eye irritation. May cause respiratory irritation
DOTInformation Hazardous
Recommended Storage Ambient

Product intended for laboratory use only