CAS RN 7440-05-3
Palladium, 0.12%, Ruthenium, 0.12%; on 3mm alumina tablets, Thermo Scientific™
CAS: 7440-05-3 Molecular Formula: Pd Molecular Weight (g/mol): 106.42 MDL Number: MFCD03613602 InChI Key: KDLHZDBZIXYQEI-UHFFFAOYSA-N Synonym: black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon PubChem CID: 23938 ChEBI: CHEBI:33363 IUPAC Name: palladium SMILES: [Pd]
Palladium on activated carbon, 10% Pd, (50% wet with water for safety), unreduced
CAS: 7440-05-3 Molecular Formula: Pd Molecular Weight (g/mol): 106.42 MDL Number: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI Key: KDLHZDBZIXYQEI-UHFFFAOYSA-N Synonym: black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon PubChem CID: 23938 ChEBI: CHEBI:33363 IUPAC Name: palladium SMILES: [Pd]
Palladium on activated carbon, 3% Pd, unreduced, Thermo Scientific Chemicals
CAS: 7440-05-3 Molecular Formula: Pd Molecular Weight (g/mol): 106.42 MDL Number: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI Key: KDLHZDBZIXYQEI-UHFFFAOYSA-N Synonym: black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon PubChem CID: 23938 ChEBI: CHEBI:33363 IUPAC Name: palladium SMILES: [Pd]
Palladium on activated carbon, 10% Pd, unreduced, dry
CAS: 7440-05-3 Molecular Formula: Pd Molecular Weight (g/mol): 106.42 MDL Number: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI Key: KDLHZDBZIXYQEI-UHFFFAOYSA-N Synonym: black,on carbon,paladio,palladium/carbon,element,charcoal,palladium, element,palladex 600,on charcoal,palladium-activated carbon PubChem CID: 23938 ChEBI: CHEBI:33363 IUPAC Name: palladium SMILES: [Pd]
Palladium standard solution, for AAS, 1 mg/ml Pd in 10 to 20% HCl
CAS: 7440-05-3 | Pd | 106.42 g/mol
Palladium ICP Standard, 10,000 ppm Pd in 10% HCl, Ricca Chemical
CAS: 7440-05-3 Molecular Formula: Pd Molecular Weight (g/mol): 106.42 InChI Key: KDLHZDBZIXYQEI-UHFFFAOYSA-N PubChem CID: 23938 ChEBI: CHEBI:33363 IUPAC Name: palladium SMILES: [Pd]
Palladium AA Standard, 1000 ppm Pd in 3% HCl, Ricca Chemical
CAS: 7440-05-3 Molecular Formula: Pd Molecular Weight (g/mol): 106.42 MDL Number: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI Key: KDLHZDBZIXYQEI-UHFFFAOYSA-N PubChem CID: 23938 ChEBI: CHEBI:33363 IUPAC Name: palladium SMILES: [Pd]
Palladium ICP Standard, 1000 ppm Pd in 3% HCl, Ricca Chemical
CAS: 7440-05-3 Molecular Formula: Pd Molecular Weight (g/mol): 106.42 MDL Number: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI Key: KDLHZDBZIXYQEI-UHFFFAOYSA-N PubChem CID: 23938 ChEBI: CHEBI:33363 IUPAC Name: palladium SMILES: [Pd]
Palladium Standard For ICP, 1000μg/mL in 10% HCl, Assurance Grade, SPEX CertiPrep™
CAS: 7440-05-3 Molecular Formula: Pd Molecular Weight (g/mol): 106.42 MDL Number: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI Key: KDLHZDBZIXYQEI-UHFFFAOYSA-N PubChem CID: 23938 ChEBI: CHEBI:33363 IUPAC Name: palladium SMILES: [Pd]
5 Percent Palladium on Activated Carbon Powder, Unreduced, Water Wet, 27 microns, BASF Catalyst, Spectrum™ Chemical
CAS: 7440-05-3 Molecular Formula: Pd Molecular Weight (g/mol): 106.42 MDL Number: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI Key: KDLHZDBZIXYQEI-UHFFFAOYSA-N IUPAC Name: palladium SMILES: [Pd]
20 Percent Palladium on Activated Carbon Powder, Unreduced, Water Wet (Pearlman Catalyst), 24 microns, BASF Catalyst, Spectrum™ Chemical
CAS: 7440-05-3 Molecular Formula: Pd Molecular Weight (g/mol): 106.42 MDL Number: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI Key: KDLHZDBZIXYQEI-UHFFFAOYSA-N IUPAC Name: palladium SMILES: [Pd]
5 Percent Palladium on Alumina Powder, Reduced, Dry, 70 microns, BASF Catalyst, Spectrum™ Chemical
CAS: 7440-05-3 Molecular Formula: Pd Molecular Weight (g/mol): 106.42 MDL Number: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI Key: KDLHZDBZIXYQEI-UHFFFAOYSA-N IUPAC Name: palladium SMILES: [Pd]
3 Percent Palladium on Activated Carbon Powder, Unreduced, Water Wet, 30 microns, BASF Catalyst, Spectrum™ Chemical
CAS: 7440-05-3 Molecular Formula: Pd Molecular Weight (g/mol): 106.42 MDL Number: MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745 InChI Key: KDLHZDBZIXYQEI-UHFFFAOYSA-N IUPAC Name: palladium SMILES: [Pd]
- 1
- 2