missing translation for 'onlineSavingsMsg'
Learn More

Palladium ICP Standard, 10,000 ppm Pd in 10% HCl, Ricca Chemical

Supplier:  Ricca Chemical Company PPD10KH100

Packaging Natural LDPE Bottle

 View more versions of this product

Catalog No. PPD10KH100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

7440-05-3 , 7647-01-0 , 7732-18-5
106.42
23938
palladium
Pd
KDLHZDBZIXYQEI-UHFFFAOYSA-N
CHEBI:33363
[Pd]

Specifications

Palladium ICP Standard
7440-05-3 , 7647-01-0 , 7732-18-5
67.0, 23.73, 0.67
1 mL = 10 mg Pd
KDLHZDBZIXYQEI-UHFFFAOYSA-N
palladium
23938
Natural LDPE Bottle
10,000ppm Pd
70.0, 24.7, 0.7
Pd
[Pd]
106.42
CHEBI:33363
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents