missing translation for 'onlineSavingsMsg'
Learn More

Palladium ICP Standard, 1000 ppm Pd in 3% HCl, Ricca Chemical

Catalog No. ppd1kh100
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Natural LDPE Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PPD1KH100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PPD1KH100 Supplier Ricca Chemical Company Supplier No. PPD1KH100
Only null left

Chemical Identifiers

CAS 7440-05-3 , 7647-01-0 , 7732-18-5
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Packaging Natural LDPE Bottle
Concentration or Composition Notes 1,000ppm Pd
CAS Min % 92.0, 5.4, 0.09
CAS Max % 95.0, 5.63, 0.1
Equivalence 1 mL = 1 mg Pd
Physical Form Liquid
Color Brown
Melting Point Approximately 0°C
Type Standard
Quantity 100 mL
Chemical Name or Material Palladium ICP Standard
Show More Show Less