missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Palatinose hydrate, 98+%

CAS: 343336-76-5 | C12H22O11 | 342.30 g/mol

$80.08 - $172.19

Identifiants chimiques

CAS 343336-76-5
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00076094
InChI Key PVXPPJIGRGXGCY-VVLSOAGWNA-N
Synonym 6-o-alpha-d-glucopyranosyl-beta-d-fructofuranose, beta-palatinose, beta-f-palatinose, alpha-d-glucopyranose-1->6-beta-d-fructofuranose, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4s,5r-3,4,5-trihydroxy-5-hydroxymethyl oxolan-2-yl methoxy oxane-3,4,5-triol
PubChem CID 52994241
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[[(2R,3S,4S)-3,4,5-trihydroxy-5-(hydroxymethyl)oxolan-2-yl]methoxy]oxane-3,4,5-triol;hydrate
SMILES OC[C@H]1O[C@H](OC[C@H]2O[C@@](O)(CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
AAJ6009114
Afficher les documents Détails de la promotion
Thermo Scientific Chemicals
J6009114
25 g
chaque for $80.08
Il en reste null
 
AAJ6009122
Afficher les documents Détails de la promotion
Thermo Scientific Chemicals
J6009122
100 g
chaque for $172.19
Il en reste null
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Identifiants chimiques

343336-76-5
342.30
PVXPPJIGRGXGCY-VVLSOAGWNA-N
52994241
OC[C@H]1O[C@H](OC[C@H]2O[C@@](O)(CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
C12H22O11
MFCD00076094
6-o-alpha-d-glucopyranosyl-beta-d-fructofuranose, beta-palatinose, beta-f-palatinose, alpha-d-glucopyranose-1->6-beta-d-fructofuranose, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4s,5r-3,4,5-trihydroxy-5-hydroxymethyl oxolan-2-yl methoxy oxane-3,4,5-triol
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[[(2R,3S,4S)-3,4,5-trihydroxy-5-(hydroxymethyl)oxolan-2-yl]methoxy]oxane-3,4,5-triol;hydrate

Spécifications

343336-76-5
White
MFCD00076094
14,5182
PVXPPJIGRGXGCY-VVLSOAGWNA-N
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[[(2R,3S,4S)-3,4,5-trihydroxy-5-(hydroxymethyl)oxolan-2-yl]methoxy]oxane-3,4,5-triol;hydrate
52994241
≥98%
Palatinose hydrate
122°C to 124°C
C12H22O11
1757215
6-o-alpha-d-glucopyranosyl-beta-d-fructofuranose, beta-palatinose, beta-f-palatinose, alpha-d-glucopyranose-1->6-beta-d-fructofuranose, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4s,5r-3,4,5-trihydroxy-5-hydroxymethyl oxolan-2-yl methoxy oxane-3,4,5-triol
OC[C@H]1O[C@H](OC[C@H]2O[C@@](O)(CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O
342.30
342.3
Powder
Videos
Sécurité et manipulation

Sécurité et manipulation

EINECSNumber : 237-282-1

RTECSNumber : LS7175420

TSCA : No

Recommended Storage : Ambient temperatures

FDS
Documents

Documents

RUO – Research Use Only