missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Palatinose hydrate, 98+%

Catalog No. AAJ6009122
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6009122 100 g
AAJ6009114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6009122 Supplier Thermo Scientific Chemicals Supplier No. J6009122

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 343336-76-5 | C12H22O11 | 342.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 343336-76-5
Molecular Formula C12H22O11
Molecular Weight (g/mol) 342.30
MDL Number MFCD00076094
InChI Key PVXPPJIGRGXGCY-VVLSOAGWNA-N
Synonym 6-o-alpha-d-glucopyranosyl-beta-d-fructofuranose, beta-palatinose, beta-f-palatinose, alpha-d-glucopyranose-1->6-beta-d-fructofuranose, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2r,3s,4s,5r-3,4,5-trihydroxy-5-hydroxymethyl oxolan-2-yl methoxy oxane-3,4,5-triol
PubChem CID 52994241
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[[(2R,3S,4S)-3,4,5-trihydroxy-5-(hydroxymethyl)oxolan-2-yl]methoxy]oxane-3,4,5-triol;hydrate
SMILES OC[C@H]1O[C@H](OC[C@H]2O[C@@](O)(CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

Specifications

Melting Point 122°C to 124°C
Color White
Quantity 100 g
Beilstein 1757215
Merck Index 14,5182
Formula Weight 342.3
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material Palatinose hydrate
EINECSNumber 237-282-1
RTECSNumber LS7175420
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only