missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Pack of 12 Wide Mouth Polypropylene Translucent Reagent Bottles with Polypropylene Caps
This pack of 12 wide-mouth translucent polypropylene bottles with polypropylene caps is leakproof, chemical-resistant, and autoclavable, ideal for easy filling, safe storage, and versatile use in lab and industrial settings.
$52.39 - $258.29
Specifications
| Color | Translucent |
|---|---|
| Material | Polypropylene |
| No. per Pack | 12 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
S000111
|
Eisco Labs
CH5451PK12 |
30 mL |
Pack of 12 Each for $52.39
|
|
|||||
|
S000112
|
Eisco Labs
CH5452PK12 |
60 mL |
Pack of 12 Each for $59.36
|
|
|||||
|
S000113
|
Eisco Labs
CH5453PK12 |
125 mL |
Pack of 12 Each for $69.82
|
|
|||||
|
S000114
|
Eisco Labs
CH5454PK12 |
250 mL |
Pack of 12 Each for $97.71
|
|
|||||
|
S000115
|
Eisco Labs
CH5456PK12 |
1 L |
Pack of 12 Each for $258.29
|
|
|||||
Description
- Pack of 12 wide mouth reagent bottles: Includes twelve translucent polypropylene bottles with wide mouth openings and polypropylene caps for convenient laboratory and industrial use.
- Autoclavable and versatile: Supports sterilization up to 135°C, making it suitable for diverse laboratory processes, including reagent storage and sample handling.
- Chemical resistant materials: Polypropylene construction provides excellent resistance to a variety of chemicals, ensuring durability and versatility for various lab applications.
- Leakproof design: Rigorously tested to prevent leaks and evaporation, guaranteeing secure transport and storage for reagents, solutions, and samples.
- Easy pouring and transportation: Wide mouth allows easy pouring and filling. Translucent design enables content visibility, while lightweight construction ensures ease of handling.
Specifications
| Translucent | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Autoclavable |
| Polypropylene | |
| Non-sterile | |
| No | |
| Reagent Bottle |