Learn More
Eisco Labs Pack of 12 Wide Mouth Polypropylene Translucent Reagent Bottles with Polypropylene Caps
This pack of 12 wide-mouth translucent polypropylene bottles with polypropylene caps is leakproof, chemical-resistant, and autoclavable, ideal for easy filling, safe storage, and versatile use in lab and industrial settings.
Supplier:  Eisco Labs CH5453PK12
 View more versions of this product
Catalog No. S000113
Only null left
Description
Description
  • Pack of 12 wide mouth reagent bottles: Includes twelve translucent polypropylene bottles with wide mouth openings and polypropylene caps for convenient laboratory and industrial use.
  • Autoclavable and versatile: Supports sterilization up to 135°C, making it suitable for diverse laboratory processes, including reagent storage and sample handling.
  • Chemical resistant materials: Polypropylene construction provides excellent resistance to a variety of chemicals, ensuring durability and versatility for various lab applications.
  • Leakproof design: Rigorously tested to prevent leaks and evaporation, guaranteeing secure transport and storage for reagents, solutions, and samples.
  • Easy pouring and transportation: Wide mouth allows easy pouring and filling. Translucent design enables content visibility, while lightweight construction ensures ease of handling.
Specifications
Chemical Identifiers
Translucent
12 Pk.
Yes
Wide
Autoclavable
Polypropylene
Non-sterile
No
Reagent Bottle
Specifications
125 mL
Translucent
Polypropylene
Non-sterile
No
98 mm
51 mm
4.22 oz.
28.8 mm
12 Pk.
Yes
Wide
Reagent Bottle
Autoclavable
Product Suggestions
Product Suggestions