missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Pack of 12 Wide Mouth Polypropylene Translucent Reagent Bottles with Polypropylene Caps
This pack of 12 wide-mouth translucent polypropylene bottles with polypropylene caps is leakproof, chemical-resistant, and autoclavable, ideal for easy filling, safe storage, and versatile use in lab and industrial settings.
Supplier: Eisco Labs CH5452PK12
Description
- Pack of 12 wide mouth reagent bottles: Includes twelve translucent polypropylene bottles with wide mouth openings and polypropylene caps for convenient laboratory and industrial use.
- Autoclavable and versatile: Supports sterilization up to 135°C, making it suitable for diverse laboratory processes, including reagent storage and sample handling.
- Chemical resistant materials: Polypropylene construction provides excellent resistance to a variety of chemicals, ensuring durability and versatility for various lab applications.
- Leakproof design: Rigorously tested to prevent leaks and evaporation, guaranteeing secure transport and storage for reagents, solutions, and samples.
- Easy pouring and transportation: Wide mouth allows easy pouring and filling. Translucent design enables content visibility, while lightweight construction ensures ease of handling.
Chemical Identifiers
| Translucent | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Autoclavable |
| Polypropylene | |
| Non-sterile | |
| No | |
| Reagent Bottle |
Specifications
| 60 mL | |
| Translucent | |
| Polypropylene | |
| Non-sterile | |
| No | |
| 85 mm | |
| 39.8 mm |
| 2 oz. | |
| 21.6 mm | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Reagent Bottle | |
| Autoclavable |