missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Pack of 12 Wide Mouth Polypropylene Translucent Reagent Bottles with Polypropylene Caps

Catalog No. S000111
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000111 30 mL
S000112 60 mL
S000113 125 mL
S000114 250 mL
S000115 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S000111 Supplier Eisco Labs Supplier No. CH5451PK12

This pack of 12 wide-mouth translucent polypropylene bottles with polypropylene caps is leakproof, chemical-resistant, and autoclavable, ideal for easy filling, safe storage, and versatile use in lab and industrial settings.

  • Pack of 12 wide mouth reagent bottles: Includes twelve translucent polypropylene bottles with wide mouth openings and polypropylene caps for convenient laboratory and industrial use.
  • Autoclavable and versatile: Supports sterilization up to 135°C, making it suitable for diverse laboratory processes, including reagent storage and sample handling.
  • Chemical resistant materials: Polypropylene construction provides excellent resistance to a variety of chemicals, ensuring durability and versatility for various lab applications.
  • Leakproof design: Rigorously tested to prevent leaks and evaporation, guaranteeing secure transport and storage for reagents, solutions, and samples.
  • Easy pouring and transportation: Wide mouth allows easy pouring and filling. Translucent design enables content visibility, while lightweight construction ensures ease of handling.

Specifications

Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Translucent
Diameter (Metric) Inner Neck 21.6 mm
Material Polypropylene
No. per Pack 12 Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide
Height (Metric) 62 mm
Product Type Reagent Bottle
Diameter (Metric) 34.8 mm
Autoclavable Autoclavable
Show More Show Less