missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Pack of 12 Wide Mouth LDPE Reagent Bottles with Polypropylene Caps

Catalog No. S000133
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
250 mL
500 mL
1 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000133 30 mL
S000134 60 mL
S000135 250 mL
S000136 500 mL
S000137 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. S000133 Supplier Eisco Labs Supplier No. CH5515PK12

Pack of 12 wide-mouth LDPE reagent bottles with polypropylene caps are non-sterile, leak-tested, and chemically resistant for safe, easy handling and storage of lab liquids in research and industrial settings.

  • Durable LDPE construction: Pack of 12 reagent bottles made from LDPE with polypropylene caps. Designed for reliable chemical resistance and safe liquid storage.
  • Wide mouth design: Wide-mouth opening ensures easy filling, pouring, and handling, ideal for laboratory and industrial applications.
  • Chemical resistance: Resistant to a variety of chemicals, making it suitable for diverse uses in laboratories, research, and industrial environments. Non-sterile and non-autoclavable, with a temperature range of -100°C to +80°C.
  • Leakproof testing: Bottles are tested for leakage under high pressure, ensuring safe storage and transportation of liquids and reagents.
  • Translucent and lightweight: Translucent material provides visibility of contents, while lightweight construction ensures ease of carrying and handling.

Specifications

Capacity (Metric) 30 mL
Capacity (English) 1 oz.
Color Translucent
Diameter (Metric) Inner Neck 21.6 mm
Material LDPE
No. per Pack 12 Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide
Height (Metric) 62 mm
Product Type Reagent Bottle
Diameter (Metric) 34.8 mm
Autoclavable Not Autoclavable
Show More Show Less