missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Pack of 12 Wide Mouth LDPE Reagent Bottles with Polypropylene Caps
Description
- Durable LDPE construction: Pack of 12 reagent bottles made from LDPE with polypropylene caps. Designed for reliable chemical resistance and safe liquid storage.
- Wide mouth design: Wide-mouth opening ensures easy filling, pouring, and handling, ideal for laboratory and industrial applications.
- Chemical resistance: Resistant to a variety of chemicals, making it suitable for diverse uses in laboratories, research, and industrial environments. Non-sterile and non-autoclavable, with a temperature range of -100°C to +80°C.
- Leakproof testing: Bottles are tested for leakage under high pressure, ensuring safe storage and transportation of liquids and reagents.
- Translucent and lightweight: Translucent material provides visibility of contents, while lightweight construction ensures ease of carrying and handling.
Specifications
Specifications
| Capacity (Metric) | 1 L |
| Color | Translucent |
| Diameter (Metric) Inner Neck | 53 mm |
| Material | LDPE |
| No. per Pack | 12 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Height (Metric) | 198 mm |
| Show More |