missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Pack of 12 Wide Mouth Amber HDPE Reagent Bottles with Polypropylene Caps
Pack of 12 non-sterile wide-mouth amber HDPE reagent bottles with polypropylene caps are UV-blocking, leak-tested, and chemically resistant for safe storage and easy handling of light-sensitive liquids in lab or industrial use.
$53.86 - $265.50
Specifications
| Color | Amber |
|---|---|
| Material | HDPE |
| No. per Pack | 12 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
S000124
|
Eisco Labs
CH5491PK12 |
30 mL |
Pack of 12 Each for $53.86
|
|
|||||
|
S000125
|
Eisco Labs
CH5492PK12 |
60 mL |
Pack of 12 Each for $61.04
|
|
|||||
|
S000126
|
Eisco Labs
CH5493PK12 |
125 mL |
Pack of 12 Each for $71.75
|
|
|||||
|
S000127
|
Eisco Labs
CH5494PK12 |
250 mL |
Pack of 12 Each for $100.46
|
|
|||||
|
S000128
|
Eisco Labs
CH5495PK12 |
500 mL |
Pack of 12 Each for $143.50
|
|
|||||
|
S000129
|
Eisco Labs
CH5496PK12 |
1 L |
Pack of 12 Each for $265.50
|
|
|||||
Description
- Pack of 12 reagent bottles: Includes 12 wide-mouth amber HDPE bottles with polypropylene caps, designed for lab storage and secure transport of light-sensitive samples.
- Light-shielding design: Amber color protects light-sensitive substances by blocking UV rays, ensuring safe storage of photosensitive materials in labs.
- Leak-proof construction: Bottles are leak-tested, offering secure containment for various chemicals and reagents.
- Chemical-resistant materials: Constructed from durable HDPE, these bottles offer excellent chemical resistance for versatile lab and industrial applications.
- Convenient and reliable: Wide neck allows easy filling and pouring.
Specifications
| Amber | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Not Autoclavable |
| HDPE | |
| Non-sterile | |
| No | |
| Reagent Bottle |