missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Pack of 12 Wide Mouth Amber HDPE Reagent Bottles with Polypropylene Caps
Description
- Pack of 12 reagent bottles: Includes 12 wide-mouth amber HDPE bottles with polypropylene caps, designed for lab storage and secure transport of light-sensitive samples.
- Light-shielding design: Amber color protects light-sensitive substances by blocking UV rays, ensuring safe storage of photosensitive materials in labs.
- Leak-proof construction: Bottles are leak-tested, offering secure containment for various chemicals and reagents.
- Chemical-resistant materials: Constructed from durable HDPE, these bottles offer excellent chemical resistance for versatile lab and industrial applications.
- Convenient and reliable: Wide neck allows easy filling and pouring.
Specifications
Specifications
| Capacity (Metric) | 60 mL |
| Capacity (English) | 2 oz. |
| Color | Amber |
| Diameter (Metric) Inner Neck | 21.6 mm |
| Material | HDPE |
| No. per Pack | 12 Pk. |
| Sterility | Non-sterile |
| Closure Included | Yes |
| Graduated | No |
| Mouth | Wide |
| Show More |