missing translation for 'onlineSavingsMsg'
Learn More

Eisco Labs Pack of 12 Wide Mouth Amber HDPE Reagent Bottles with Polypropylene Caps

Catalog No. S000129
Change view
Click to view available options
Capacity (Metric):
30 mL
60 mL
125 mL
250 mL
500 mL
1 L
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S000129 1 L
S000124 30 mL
S000125 60 mL
S000126 125 mL
S000127 250 mL
S000128 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S000129 Supplier Eisco Labs Supplier No. CH5496PK12

Pack of 12 non-sterile wide-mouth amber HDPE reagent bottles with polypropylene caps are UV-blocking, leak-tested, and chemically resistant for safe storage and easy handling of light-sensitive liquids in lab or industrial use.

  • Pack of 12 reagent bottles: Includes 12 wide-mouth amber HDPE bottles with polypropylene caps, designed for lab storage and secure transport of light-sensitive samples.
  • Light-shielding design: Amber color protects light-sensitive substances by blocking UV rays, ensuring safe storage of photosensitive materials in labs.
  • Leak-proof construction: Bottles are leak-tested, offering secure containment for various chemicals and reagents.
  • Chemical-resistant materials: Constructed from durable HDPE, these bottles offer excellent chemical resistance for versatile lab and industrial applications.
  • Convenient and reliable: Wide neck allows easy filling and pouring.

Specifications

Capacity (Metric) 1 L
Color Amber
Diameter (Metric) Inner Neck 53 mm
Material HDPE
No. per Pack 12 Pk.
Sterility Non-sterile
Closure Included Yes
Graduated No
Mouth Wide
Height (Metric) 198 mm
Diameter (Metric) 93.5 mm
Autoclavable Not Autoclavable
Capacity (English) 33.8 oz.
Product Type Reagent Bottle
Show More Show Less