missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Pack of 12 Wide Mouth Amber HDPE Reagent Bottles with Polypropylene Caps
Pack of 12 non-sterile wide-mouth amber HDPE reagent bottles with polypropylene caps are UV-blocking, leak-tested, and chemically resistant for safe storage and easy handling of light-sensitive liquids in lab or industrial use.
Supplier: Eisco Labs CH5495PK12
Description
- Pack of 12 reagent bottles: Includes 12 wide-mouth amber HDPE bottles with polypropylene caps, designed for lab storage and secure transport of light-sensitive samples.
- Light-shielding design: Amber color protects light-sensitive substances by blocking UV rays, ensuring safe storage of photosensitive materials in labs.
- Leak-proof construction: Bottles are leak-tested, offering secure containment for various chemicals and reagents.
- Chemical-resistant materials: Constructed from durable HDPE, these bottles offer excellent chemical resistance for versatile lab and industrial applications.
- Convenient and reliable: Wide neck allows easy filling and pouring.
Chemical Identifiers
| Amber | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Not Autoclavable |
| HDPE | |
| Non-sterile | |
| No | |
| Reagent Bottle |
Specifications
| 500 mL | |
| Amber | |
| HDPE | |
| Non-sterile | |
| No | |
| 167 mm | |
| 73.7 mm |
| 17 oz. | |
| 43.7 mm | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Reagent Bottle | |
| Not Autoclavable |