missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Pack of 12 Sterile Wide Mouth Amber HDPE Reagent Bottles with Polypropylene Caps
Pack of 12 sterile wide-mouth amber HDPE reagent bottles with polypropylene caps areUV-protective, leak-tested, and chemically resistant, ideal for safe storage and transfer of light-sensitive liquids in lab and industrial settings.
$62.82 - $122.18
Specifications
| Color | Amber |
|---|---|
| Material | HDPE |
| No. per Pack | 12 Pk. |
| Sterility | Sterile |
| Closure Included | Yes |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity & Availability | |||||
|
S000130
|
Eisco Labs
CH5501PK12 |
30 mL |
Pack of 12 Each for $62.82
|
|
|||||
|
S000131
|
Eisco Labs
CH5503PK12 |
125 mL |
Pack of 12 Each for $87.29
|
|
|||||
|
S000132
|
Eisco Labs
CH5504PK12 |
250 mL |
Pack of 12 Each for $122.18
|
|
|||||
Description
- Amber reagent bottles: Includes 12 sterile wide-mouth HDPE bottles with polypropylene caps, designed for laboratory sample storage and secure transportation.
- UV light protection: Amber color shields light-sensitive samples from harmful UV rays, ideal for protecting reagents, solutions, and light-sensitive substances.
- Leak-proof design: Bottles are rigorously leak-proof tested to ensure safe storage and transport of chemicals, liquids, or reagents in laboratory and industrial settings.
- Chemical-resistant materials: Manufactured from durable HDPE with excellent chemical resistance, suitable for diverse applications in laboratories and industries.
- Easy to handle and transport: Wide-mouth design allows easy filling and pouring.
Specifications
| Amber | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Not Autoclavable |
| HDPE | |
| Sterile | |
| No | |
| Reagent Bottle |