missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eisco Labs Pack of 12 Sterile Wide Mouth Amber HDPE Reagent Bottles with Polypropylene Caps
Pack of 12 sterile wide-mouth amber HDPE reagent bottles with polypropylene caps areUV-protective, leak-tested, and chemically resistant, ideal for safe storage and transfer of light-sensitive liquids in lab and industrial settings.
Supplier: Eisco Labs CH5503PK12
Description
- Amber reagent bottles: Includes 12 sterile wide-mouth HDPE bottles with polypropylene caps, designed for laboratory sample storage and secure transportation.
- UV light protection: Amber color shields light-sensitive samples from harmful UV rays, ideal for protecting reagents, solutions, and light-sensitive substances.
- Leak-proof design: Bottles are rigorously leak-proof tested to ensure safe storage and transport of chemicals, liquids, or reagents in laboratory and industrial settings.
- Chemical-resistant materials: Manufactured from durable HDPE with excellent chemical resistance, suitable for diverse applications in laboratories and industries.
- Easy to handle and transport: Wide-mouth design allows easy filling and pouring.
Chemical Identifiers
| Amber | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Not Autoclavable |
| HDPE | |
| Sterile | |
| No | |
| Reagent Bottle |
Specifications
| 125 mL | |
| Amber | |
| HDPE | |
| Sterile | |
| No | |
| 98 mm | |
| 51 mm |
| 4.22 oz. | |
| 28.8 mm | |
| 12 Pk. | |
| Yes | |
| Wide | |
| Reagent Bottle | |
| Not Autoclavable |